Description
- Prepacked with Sephacryl™ S-200 HR medium pre-equilibrated in TE buffer
- Percent sample recovery: >80%
- Tested in nickase, single- and double-stranded exonuclease and RNase assays
Specifications
Specifications
| Format | Spin Column |
| For Use With (Application) | Rapid purification of labeled single-stranded or double-stranded DNA fragments <50 bases in length in a volume of 25 to 100μL using spin-column chromatography |
| Product Type | Column |
| Quantity | Each |