Description
Enzymes optimized to work together for high efficiency removal of unincorporated primers and dNTPs. Enzymes provided in two separate tubes, just two simple pipetting steps are needed to prepare the reaction.
- Fast 30-minute protocol
- Scalable for different reaction sizes
- No loss of PCR product
- Easy to automate
- Complete heat inactivation of the enzymes within 15 minutes
Specifications
Specifications
| Description | Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes. |
| For Use With (Application) | Optimized to purify DNA and sequencing set up reactions quickly, efficiently and reliably |
| Includes | One tube of Alkaline Phosphatase and Exonuclease I |
| No. of Reactions | 500 |
| Product Type | illustra ExoProStar |
Safety and Handling
Product Identifier
- illustra
- EUH210-Safety data sheet available on request.
- UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.