missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24926-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. ILK is a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction.Specifications
ILK | |
Polyclonal | |
Unconjugated | |
ILK | |
ILK, ILK 1, ILK 2, ILK1, ILK2, integrin linked kinase, Integrin linked protein kinase, P59, p59ILK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16202, 3611 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O55222, Q13418 | |
ILK | |
ILK Fusion Protein Ag21494 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |