Learn More
Abnova™ IL9R (Human) Recombinant Protein
Product Code. p-7164381
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. Quantity unitSize
16116811 10 μg 10µg
16126811 25 μg 25µg
2 options
Product Code. 16116811
Brand: Abnova™ H00003581P01.10ug
Human IL9R full-length ORF ( AAH51337, 1 a.a. - 332 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MHLGSNCCKNGQTLLQRTCHGVSCCGWWFQAARSILGKGPSAQSLAGWTLESEALRRDMGTWLLACICICTCVCLGVSVTGEGQGPRSRTFTCLTNNILRIDCHWSAPELGQGSSPWLLFTRLLAAHISASCGAVSAPSCCHLRQCSCHLTISPSLSTTACLGGSRSAWWTRSTCPGDTSNISSGHCILTWSISPALEPMTTLLSYELAFKKQEEAWEQAQHRDHIVGVTWLILEAFELDPGFIHEARLRVQMATLEDDVVEEERYTGQWSEWSQPVCFQAPQRQGPLIPPWGWPGNTLVAVSIFLLLTGPTYLLFKLSPRLGWGPTGPVCC
Specifications
Accession Number AAH51337
Gene ID (Entrez) 3581
Name interleukin 9 receptor
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CD129
Gene Symbol IL9R
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer