missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL3RA Monoclonal Antibody (OTI10D4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813796
Additional Details : Weight : 0.05000kg
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
IL3RA Monoclonal antibody specifically detects IL3RA in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
IL3RA | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P26951 | |
Mouse | |
Protein A/G | |
RUO | |
3563 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
OTI10D4 | |
Unconjugated | |
IL3RA | |
CD123; CD123 antigen; CDw123; Cyrl; cytokine receptor-like protein CYRL; hIL-3Ra; Il-3 alpha subunit; IL-3 receptor alpha chain; IL-3 receptor alpha SP2 isoform; IL-3 receptor subunit alpha; IL3R; IL-3R subunit alpha; Il3ra; IL-3RA; IL-3R-alpha; IL3RAY; IL3RX; IL3RY; interleukin 3 receptor subunit alpha; interleukin 3 receptor, alpha; interleukin 3 receptor, alpha (low affinity); interleukin 3 receptor, alpha chain; interleukin-3 receptor class II alpha chain; Interleukin-3 receptor subunit alpha; Sut-1 | |
Full length human recombinant protein of human IL3RA (NP_002174) produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |