Learn More
IL3RA Monoclonal Antibody (OTI10D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813795
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
IL3RA Monoclonal antibody specifically detects IL3RA in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| IL3RA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P26951 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3563 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| OTI10D1 | |
| Unconjugated | |
| IL3RA | |
| CD123; CD123 antigen; CDw123; Cyrl; cytokine receptor-like protein CYRL; hIL-3Ra; Il-3 alpha subunit; IL-3 receptor alpha chain; IL-3 receptor alpha SP2 isoform; IL-3 receptor subunit alpha; IL3R; IL-3R subunit alpha; Il3ra; IL-3RA; IL-3R-alpha; IL3RAY; IL3RX; IL3RY; interleukin 3 receptor subunit alpha; interleukin 3 receptor, alpha; interleukin 3 receptor, alpha (low affinity); interleukin 3 receptor, alpha chain; interleukin-3 receptor class II alpha chain; Interleukin-3 receptor subunit alpha; Sut-1 | |
| Full length human recombinant protein of human IL3RA (NP_002174) produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.