missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IL1F7 Recombinant Protein
Brand: Abnova™ H00027178-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH20637 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
50.1 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MSFVGENSGVKMGSEDWEKDEPQCCLEDPAVSPLEPGPSLPAMNFVHTSPKVKNLNPKKFSIHDQDHKVLVLDSGNLIAVPDKNYIRPEIFFALASSLSSASAEKGSPILLGVSKGEFCLYCDKDKGQSHPSLQLKKEKLMKLAAQKESARRPFIFYRAQVGSWNMLESAAHPGWFICTSCNCNEPVGVTDKFENRKHIEFSFQPVCKAEMSPSEVSD | |
FIL1/FIL1(ZETA)/FIL1Z/IL-1F7/IL-1H4/IL-1RP1/IL1H4/IL1RP1 | |
IL1F7 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
27178 | |
IL1F7 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IL1F7 | |
Human | |
Recombinant | |
Solution |