missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL12RB1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13287-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a type I transmembrane protein that belongs to the hemopoietin receptor superfamily. This protein binds to interleukine 12 with a low affinity, and is thought to be a part of IL12 receptor complex. This protein forms a disulfide-linked oligomer, which is required for its IL12 binding activity. The coexpression of this and IL12RB2 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The lack of expression of this gene was found to result in the immunodeficiency of patients with severe mycobacterial and Salmonella infections. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.Specifications
IL12RB1 | |
Polyclonal | |
Unconjugated | |
IL12RB1 | |
CD212, IL 12 receptor beta component, IL 12 receptor subunit beta 1, IL 12R beta 1, IL 12R BETA1, IL 12R subunit beta 1, IL 12RB1, IL-12 R beta 1, IL12R, IL12RB, IL12RB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3594 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P42701 | |
IL12RB1 | |
IL12RB1 Fusion Protein Ag3983 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |