missing translation for 'onlineSavingsMsg'
Learn More

IL11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55169-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16886645

  • 4593.76 NOK / 150µL
Estimated Shipment: 17-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL.
Specifications

Specifications

IL11
Polyclonal
Unconjugated
IL11
Adipogenesis inhibitory factor, AGIF, IL 11, IL11, interleukin 11, Oprelvekin
Rabbit
Antigen Affinity Chromatography
RUO
16156, 3589
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P20809, P47873
IL11
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers