missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55169-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL.Specifications
IL11 | |
Polyclonal | |
Unconjugated | |
IL11 | |
Adipogenesis inhibitory factor, AGIF, IL 11, IL11, interleukin 11, Oprelvekin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16156, 3589 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P20809, P47873 | |
IL11 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |