missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-9 Mouse anti-Human, CoraLite 594, Clone: 1F3A11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66144-20UL
This item is not returnable.
View return policy
Description
IL-9 Monoclonal antibody apecifically detetects IL-9 antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| IL-9 | |
| Monoclonal | |
| 1000 ug/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| IL9 | |
| IL-9 fusion protein Ag21435 | |
| 20 μL | |
| 3578 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG2b |
| Immunofluorescence | |
| 1F3A11 | |
| CoraLite 594 | |
| BC066284 | |
| Mouse | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction