missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-7R alpha/CD127 Antibody (40131) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB306JF585
This item is not returnable.
View return policy
Description
IL-7R alpha/CD127 Monoclonal antibody specifically detects IL-7R alpha/CD127 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| IL-7R alpha/CD127 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD127 antigen, CD127ILRA, CDW127, IL-7 receptor subunit alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
| Mouse myeloma cell line NS0-derived recombinant human IL-7 R alpha , Extracellular domain | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
| 3575 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Western Blot, CyTOF | |
| 40131 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction