missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-5R alpha/CD125 Antibody (1036601) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2531JF585
This item is not returnable.
View return policy
Description
IL-5R alpha/CD125 Monoclonal antibody specifically detects IL-5R alpha/CD125 in Human samples. It is validated for Flow Cytometry
Specifications
| IL-5R alpha/CD125 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD125 antigen, CDw125CD125, HSIL5R3, IL-5 receptor subunit alpha, IL5R, IL-5R subunit alpha, IL-5RA, IL-5R-alpha, interleukin 5 receptor type 3, interleukin 5 receptor, alpha, interleukin-5 receptor alpha chain, interleukin-5 receptor alpha subunit, interleukin-5 receptor subunit alpha, MGC26560 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-5 R alpha /CD125, Asp21-Arg335, Accession # O08665 | |
| 0.1 mL | |
| Immunology | |
| 3568 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 1036601 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction