missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-37 Monoclonal Antibody (K92047_1A9), Invitrogen™

Product Code. 30201776
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
Product Code. Quantity unitSize
30201776 200 μL 200µg
30204746 50 μL 50µg
2 options
This item is not returnable. View return policy

Product Code. 30201776

Brand: Invitrogen™ MA556097

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: KVKNLNPKKF SIHDQDHKVL VLDSGNLIAV PDKNYIRPEI FFALASSLSS ASAEKGSPIL LGVSKGEFCL YCDKDKGQSH PSLQLKKEKL

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). This cytokine also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18. This gene along with eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Five alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-37
Applications Western Blot
Classification Monoclonal
Clone K92047_1A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene IL37
Gene Accession No. Q9NZH6
Gene Alias FIL1; FIL1 zeta; FIL1(ZETA); FIL1Z; IL-1 zeta; IL1F7; IL-1F7; IL1F7 (canonical product IL-1F7b); IL-1F7b (IL-1H4, IL-1H, IL-1RP1); IL-1H; IL1H4; IL-1H4; IL1RP1; IL-1RP1; IL-1X; IL-1X protein; IL37; IL-37; ILN; Interleukin; interleukin 1 family member 7; interleukin 1, zeta; interleukin 37; Interleukin-1 family member 7; Interleukin-1 homolog 4; interleukin-1 superfamily z; Interleukin-1 zeta; Interleukin-1-related protein; interleukin-23; Interleukin37; Interleukin-37
Gene Symbols IL37
Host Species Mouse
Immunogen Recombinant human IL37 (immunogen range is 51-140).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27178
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.