Learn More
IL-23R Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27163-1-AP-150UL
This item is not returnable.
View return policy
Description
The IL-23R antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human IL-23R. This antibody recognizes human, mouse antigen. The IL-23R antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| IL-23R | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 149233 | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |