Learn More
IL-20 R beta/FNDC6 Antibody (20RNTC) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-00466JF635
Product Code. 30171610
  • 5090.00 NOK / 0.10mL
Description
IL-20 R beta/FNDC6 Monoclonal antibody specifically detects IL-20 R beta/FNDC6 in Human,Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-20 R beta/FNDC6
Monoclonal
Janelia Fluor 635
FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923
The immunogen for this antibody is FNDC6 (IL-20R2).
0.1 mL
Primary
Human, Mouse
Purified
Flow Cytometry, Immunocytochemistry/Immunofluorescence
20RNTC
50mM Sodium Borate
Rat
Protein A or G purified
RUO
53833
Store at 4°C in the dark.
IgG2a κ