Learn More
IL-20 R beta/FNDC6 Antibody (20RNTC) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-00466JF635
This item is not returnable.
View return policy
Description
IL-20 R beta/FNDC6 Monoclonal antibody specifically detects IL-20 R beta/FNDC6 in Human,Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry/ Immunofluorescence
Specifications
| IL-20 R beta/FNDC6 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923 | |
| The immunogen for this antibody is FNDC6 (IL-20R2). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| 20RNTC | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 53833 | |
| Store at 4°C in the dark. | |
| IgG2a κ |