missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-18/IL-1F4 Antibody (74801) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB646JF525
6243.25 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Monoclonal antibody specifically detects IL-18/IL-1F4 in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF,Gene Knock-Out
Specifications
| IL-18/IL-1F4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
| E. coli-derived recombinant human IL-18/IL-1F4, Accession # Q14116 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF, Gene Knock-Out | |
| 74801 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.5 mL | |
| Angiogenesis, Apoptosis, Cancer, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Endocrinology, Immunology, Innate Immunity | |
| 3606 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction