missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17F Antibody (4H1629.1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27327F
This item is not returnable.
View return policy
Description
IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IL-17F | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| IL17F | |
| Full-length recombinant MRkIPkVGHT FFQkPESCPP VPGGSMkLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGkEDISMN SVPIQQETLV VRRkHQGCSV SFQLEkVLVT VGCTCVTPVI HHVQ | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research | |
| 112744 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4H1629.1 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Cytokine ML-1, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction