missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IL-15 Antibody (ordesekimab), Novus Biologicals™

Product Code. 30047133 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
Product Code. Quantity unitSize
30047133 50 μg 50µg
30046967 1 mg 1mg
30045611 100 μg 100µg
3 options
This item is not returnable. View return policy

Product Code. 30047133

Brand: Novus Biologicals™ NBP32796650ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-15
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone ordesekimab
Conjugate Unconjugated
Formulation Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Gene Accession No. P40933
Gene Alias IL-15MGC9721, interleukin 15, interleukin-15
Host Species Human
Immunogen IL-15
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3600
Reconstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Target Species Human
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.