Learn More
Novus Biologicals™ IL-15 Antibody (ordesekimab), Novus Biologicals™
Product Code. 30046967 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
100 μg
50 μg
missing translation for 'unitSize'
100µg
1mg
50µg
Artikelnummer. Quantity unitSize
30046967 1 mg 1mg
30045611 100 μg 100µg
30047133 50 μg 50µg
3 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30046967
missing translation for 'mfr': Novus Biologicals™ NBP3279661mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Human Monoclonal Antibody
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen IL-15
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone ordesekimab
Conjugate Unconjugated
Formulation Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Gene Accession No. P40933
Gene Alias IL-15MGC9721, interleukin 15, interleukin-15
Host Species Human
Immunogen IL-15
Purification Method Protein A purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3600
Reconstitution Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Target Species Human
Content And Storage Store at -20°C in powder form. Store at -80°C once reconstituted.
Form Purified
Isotype IgG1
Mehr anzeigen Weniger anzeigen