missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-12 p70 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Product Code. 17846547
Change view
Click to view available options
Quantity:
2 x 500 μg
25 μg
50 μg
500 μg
Unit Size:
1mg
25µg
500µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17846547 25 μg 25µg
17886397 2 x 500 μg 1mg
17856567 50 μg 50µg
30054526 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17846547 Supplier PeproTech Supplier No. 500P154HGBT25UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

AA Seq: p35 Subunit: RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNAS. p40 Subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS Preparation: Produced from sera of goats immunized with highly pure Recombinant Human IL-12. Anti-Human IL-12-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-12 by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human IL-12 (500-P154HG) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human IL-12. Western Blot: To detect Human IL-12 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/ml. When used in conjunction with compatible development reagents the detection limit for Recombinant Human IL-12 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-12 p70
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS with no preservative
Gene IL12A
Gene Accession No. P29459, P29460
Gene Alias 20C2; CLMF; CLMF p35; CLMF p40; CLMF2; cytokine; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12p70; Il12; IL-12; Il-12 p35; IL-12 p75; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il-12; cytokine; IL12A; IL-12A; IL12A/B; IL12AB; IL12B; Il-12b; IL12P35; IL-12p35; Il12p40; Il-12p40; IL35 subunit; ILN; IMD28; IMD29; Interleukin; interleukin 12 35 kDa subunit; interleukin 12 35kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 40kDa subunit; interleukin 12 p35 subunit; interleukin 12 p40 subunit precursor; interleukin 12 polypeptide 35; interleukin 12, p35; interleukin 12, p40; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin 12p35 subunit precursor; interleukin-12 alpha chain; interleukin-12 alpha p40 subunit; interleukin-12 beta chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 p35 subunit precursor; interleukin-12 p40; interleukin-12 p40 subunit; interleukin-12 p40 subunit precursor; Interleukin-12 p70; interleukin-12 subunit alpha; Interleukin-12 subunit beta; interleukin-23 p40 subunit; Ll12a; MGC151228; MGC151232; natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; P35; p40
Gene Symbols IL12A, Il12b
Host Species Goat
Immunogen HEK293 cells-derived Recombinant Human IL-12 p70.
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3592, 3593
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.