missing translation for 'onlineSavingsMsg'
Learn More

IKBKG Mouse anti-Human, Clone: 1F2E2, Proteintech

Product Code. 16888355 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16888355 150 μL 150µL
16898355 20 μL 20µL
2 options
This item is not returnable. View return policy

Product Code. 16888355

Brand: Proteintech 664601IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IKK gamma is the regulatory subunit of the inhibitor of the kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome.

Specifications

Antigen IKBKG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1F2E2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene IKBKG
Gene Accession No. Q9Y6K9
Gene Alias AMCBX1, FIP 3, FIP3, Fip3p, I kappa B kinase subunit gamma, IkB kinase subunit gamma, IKBKG, IKK gamma, IKKAP1, IKKG, IP, IP1, IP2, IPD2, NEMO, NF kappa B essential modifier, NF kappa B essential modulator
Gene Symbols IKBKG
Host Species Mouse
Immunogen IKBKG Fusion Protein Ag13358
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8517
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.