missing translation for 'onlineSavingsMsg'
Learn More

IKBKG Mouse anti-Human, Clone: 1F2E2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66460-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16898355

  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IKK gamma is the regulatory subunit of the inhibitor of the kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome.
Specifications

Specifications

IKBKG
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y6K9
IKBKG
IKBKG Fusion Protein Ag13358
20 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
1F2E2
Unconjugated
IKBKG
AMCBX1, FIP 3, FIP3, Fip3p, I kappa B kinase subunit gamma, IkB kinase subunit gamma, IKBKG, IKK gamma, IKKAP1, IKKG, IP, IP1, IP2, IPD2, NEMO, NF kappa B essential modifier, NF kappa B essential modulator
Mouse
Protein G
RUO
8517
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers