missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IkB beta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12660-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The NF-kB/Rel transcription factors are present in the cytosol in an inactive state, complexed with the inhibitory IkB proteins. Most agents that activate NF-kB do so through a common pathway based on phosphorylation-induced, proteasome-mediated degradation of IkB. The key regulatory step in this pathway involves activation of a high molecular weight IkB kinase (IKK) complex whose catalysis is generally carried out by three tightly associated IKK subunits. IKK alpha and IKK beta serve as the catalytic subunits of the kinase and IKK gamma serves as the regulatory subunit. Activation of IKK depends upon phosphorylation at Ser177 and Ser181 in the activation loop of IKK beta (Ser176 and Ser180 in IKK alpha), which causes conformational changes, resulting in kinase activation.Specifications
IkB beta | |
Polyclonal | |
Unconjugated | |
NFKBIB | |
I kappa B beta, IkappaBbeta, IkB B, IkB beta, IKBB, NF kappa B inhibitor beta, NF kappa BIB, NFKBIB, TR interacting protein 9, TRIP 9, TRIP9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18036, 4793, 81525 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15653, Q60778, Q9JIA3 | |
Nfkbib | |
IkB beta Fusion Protein Ag3340 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |