Description
- Round one-hand attachment with rubber insert (continuous / touch operation)
- 88mm dia.
Specifications
Specifications
| Product Type | One hand attachment |
| Diameter (Metric) | 8 mm |
| For Use With (Equipment) | IKA Vortex 3 Mixer |