Learn More
IGLV6-57 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18344-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16888424
  • 4593.76 NOK / 150µL
Estimated Shipment: 23-05-2024
Description
The IGLV6-57 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human IGLV6-57. This antibody recognizes human antigen. The IGLV6-57 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
IGLV6-57
Polyclonal
Unconjugated
150 μL
28778
IgG
Western Blot, ELISA
427 μg/mL