Learn More
IgG3 Mouse, Unconjugated, Clone: MG3-35, Isotype Control, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand:  Stemcell Technologies 60073
Additional Details : Weight : 0.00110kg
 View more versions of this product
Product Code. 17108871
  • 2651.09 NOK / 500µg
Estimated Shipment: 12-10-2023
Description
  • The MG3-35 antibody (IgG3, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
  • Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
  • If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
  • The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
  • This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Specifications
Specifications
IgG3
Monoclonal
Unconjugated
500 μg
IgG3 κ
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
MG3-35
Mouse
Isotype Control (Primary)
Other