Learn More
IgG2b Mouse, Unconjugated, Clone: 11B8C4, Isotype Control, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66360-3-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The Mouse IgG2b isotype control antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of Mouse IgG2b isotype control. This antibody recognizes Megathura crenulata antigen. The Mouse IgG2b isotype control antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
IgG2b | |
11B8C4 | |
Unconjugated | |
150 μL | |
M. crenulata |
ChIP Assay, Flow Cytometry | |
1000 μg/mL | |
Mouse | |
Isotype Control (Primary) | |
IgG2b |