Learn More
IgG2b Mouse, PE, Clone: MPC-11, Isotype Control, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech PE-65128-100UG
Additional Details : Weight : 0.00010kg
Description
The IgG2b Isotype Control antibody from Proteintech is a mouse monoclonal antibody generated with n/a of mouse IgG2b Isotype Control. This antibody recognizes antigen. The IgG2b Isotype Control antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
IgG2b | |
MPC-11 | |
PE | |
100 μg | |
16016 |
Flow Cytometry | |
0.2 mg/mL | |
Mouse | |
Isotype Control (Primary) | |
IgG2b |