Learn More
IgG2a Mouse, CoraLite 594, Clone: 11A1B2, Isotype Control, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66360-2-100UL
Additional Details : Weight : 0.00020kg
Description
The Mouse IgG2a isotype control antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of Mouse IgG2a isotype control. This antibody recognizes Megathura crenulata antigen. The Mouse IgG2a isotype control antibody has been validated for the following applications: FC analysis. This antibody is coralite™594.
Specifications
IgG2a | |
Monoclonal | |
CoraLite 594 | |
100 μL | |
M. crenulata |
Flow Cytometry | |
11A1B2 | |
Mouse | |
Primary | |
IgG2a |