Learn More
IgG1 Mouse, Unconjugated, Clone: 1F8D3, Isotype Control, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66360-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The Mouse IgG1 isotype control antibody from Proteintech is a mouse monoclonal antibody generated with peptide of Mouse IgG1 isotype control. This antibody recognizes Megathura crenulata antigen. The Mouse IgG1 isotype control antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
IgG1 | |
1F8D3 | |
Unconjugated | |
150 μL | |
M. crenulata |
Western Blot, Immunoprecipitation, Immunohistochemistry, Flow Cytometry | |
1000 μg/mL | |
Mouse | |
Isotype Control (Primary) | |
IgG1 |