Learn More
IgG1 Mouse, Unconjugated, Clone: 1F8D3, Isotype Control, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66360-1-IG-150UL
Additional Details : Weight : 0.00010kg
Product Code. 16838165
  • 2175.99 NOK / 150µL
Estimated Shipment: 03-06-2024
Description
The Mouse IgG1 isotype control antibody from Proteintech is a mouse monoclonal antibody generated with peptide of Mouse IgG1 isotype control. This antibody recognizes Megathura crenulata antigen. The Mouse IgG1 isotype control antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
Specifications
IgG1
1F8D3
Unconjugated
150 μL
M. crenulata
Western Blot, Immunoprecipitation, Immunohistochemistry, Flow Cytometry
1000 μg/mL
Mouse
Isotype Control (Primary)
IgG1