Learn More
IgG Mouse anti-Bovine, Unconjugated, Clone: 1C5C5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67615-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The Bovine IgG antibody from Proteintech is a mouse monoclonal antibody generated with peptide of bovine Bovine IgG. This antibody recognizes bovine antigen. The Bovine IgG antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
IgG | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
IgG2a |
Western Blot, ELISA | |
1C5C5 | |
Unconjugated | |
150 μL | |
Bovine |