Learn More
IgG Heavy Chain Mouse anti-Rabbit, Unconjugated, Clone: 1C9A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66467-1-IG-150UL
Description
The Heavy chain of Rabbit IgG antibody from Proteintech is a mouse monoclonal antibody generated with protein of rabbit Heavy chain of Rabbit IgG. This antibody recognizes rabbit antigen. The Heavy chain of Rabbit IgG antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IgG Heavy Chain | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| IgG1 |
| Western Blot, ELISA | |
| 1C9A5 | |
| Unconjugated | |
| 150 μL | |
| Rabbit |