Learn More
IgG(H+L) Donkey anti-Rabbit, FITC, Secondary Antibody, Proteintech
Product Code. 17630337 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
200 μL
500 μL
Unit Size:
200µL
500µL
Product Code. Quantity unitSize
17630337 200 μL 200µL
17640337 500 μL 500µL
2 options
Product Code. 17630337
Brand: Proteintech SA000038200UL
Donkey anti-Rabbit Affinipure Secondary Antibody
Specifications
Antigen IgG(H+L)
Applications Immunofluorescence, Flow Cytometry
Conjugate FITC
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Donkey
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 200 μL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid