Learn More
IgG(H+L) Donkey anti-Mouse, HRP, Secondary Antibody, Proteintech
Donkey anti-Mouse Affinipure Secondary Antibody
Brand: Proteintech SA00001-8-1ML
Additional Details : Weight : 0.00010kg
Specifications
IgG(H+L) | |
HRP | |
Donkey | |
1 mL | |
Mouse | |
Liquid |
ELISA, Western Blot | |
0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. | |
Secondary | |
Store at -20°C. Stable for one year after shipment. |