Learn More
IgG(H+L) Donkey anti-Human, Biotin, Secondary Antibody, Proteintech
Donkey anti-Human Affinipure Secondary Antibody
589.77 NOK - 2111.37 NOK
Specifications
| Antigen | IgG(H+L) |
|---|---|
| Applications | ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry |
| Conjugate | Biotin |
| Form | Liquid |
| Host Species | Donkey |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17690337
|
Proteintech
SA00004-10-200UL |
200 μL |
589.77 NOK
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17680337
|
Proteintech
SA00004-10-1ML |
1 mL |
2111.37 NOK
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| IgG(H+L) | |
| Biotin | |
| Donkey | |
| 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
| The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. |
| ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Liquid | |
| Human | |
| Secondary | |
| Store at -20°C. Stable for one year after shipment. |