missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgD Monoclonal Antibody (IA6-2), NovaFluor™ Blue 610-30S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1626.00 NOK - 3610.00 NOK
Specifications
| Antigen | IgD |
|---|---|
| Clone | IA6-2 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.Specifications
| IgD | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| IGHD; immunoglobulin heavy constant delta | |
| IGHD | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| IGHD |
| IA6-2 | |
| Flow Cytometry | |
| NovaFluor Blue 610-30S | |
| Mouse | |
| Human | |
| P01880 | |
| 3495 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title