missing translation for 'onlineSavingsMsg'
Learn More

IFT140 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16857354 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16857354 20 μL 20µL
16847354 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16857354

Brand: Proteintech 174601AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFT140 contains 9 TPR repeats and 5 WD repeats. The function of the IFT140 protein remains unknown.

Specifications

Antigen IFT140
Applications Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene IFT140
Gene Accession No. E9PY46, Q96RY7
Gene Alias c305C8.4, c380F5.1, gs114, IFT140, KIAA0590, WDTC2
Gene Symbols Ift140
Host Species Rabbit
Immunogen IFT140 Fusion Protein Ag11119
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100362124, 106633, 9742
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.