Learn More
IFNγ Rat, anti-Mouse, APC, Clone: XMG1.2, STEMCELL Technologies
Rat Monoclonal Antibody
Brand: Stemcell Technologies 100-0278
This item is not returnable.
View return policy
Description
- The XMG1.2 antibody reacts with mouse interferon-gamma (IFN-γ), a glycosylated type II interferon comprising a homodimer of ∼20 kDa subunits in its active form
- IFN-γ is produced primarily by activated T lymphocytes and natural killer (NK) cells in response to antigens, including alloantigens, mitogens, and enterotoxins
- IFN-γ binds with high affinity to the α subunit of the IFN-γ receptor, resulting in the ligand-induced assembly of a tetrameric α2β2 receptor complex capable of mediating the cellular response to the cytokine through activation of the JAK-STAT pathway
- The IFN-γ receptor is expressed on the surface of all nucleated cells
- IFN-γ has important functions in a wide range of biological processes, including macrophage activation, mediating the immune response to viruses and bacteria, and modulating cellular proliferation and apoptosis
- The XMG1.2 antibody is suitable for detection of intracellular IFN-γ by methods such as flow cytometry, when paired with an appropriate secondary antibody
- The XMG1.2 antibody is also widely used in various assays to neutralize the functional activity of IFN-γ
Specifications
| IFNγ | |
| Monoclonal | |
| APC | |
| 100 μg | |
| Mouse |
| ELISA, Flow Cytometry | |
| XMG1.2 | |
| Rat | |
| Primary | |
| Dendritic Cells, Macrophages, NK Cells, T Cells |