missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma R1/CD119 Mouse anti-Human, Alexa Fluor 700, Clone: 92101, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB6731N-100UG
This item is not returnable.
View return policy
Description
IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Human samples. It is validated for Flow Cytometry.Specifications
| IFN-gamma R1/CD119 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P15260.1 | |
| IFNGR1 | |
| Mouse myeloma cell line NS0-derived recombinant human IFN-gamma R1 Glu18-Gly245 Accession # P15260.1 | |
| RUO | |
| 3459 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 92101 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFNgammaR1, IFN-gamma-R1, IFNGR, IFN-gR1, IFNGR1, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human IFN-gamma R1 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant mouse IFN-gamma R1, recombinant human (rh) IFN-gamma R2, or rhIL-10 R beta . | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only (RUO)