missing translation for 'onlineSavingsMsg'
Learn More

IFITM1 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Product Code. 16898993 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16898993 20 μL 20µL
16888993 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16898993

Brand: Proteintech 117273AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFITM1 (Interferon inducible transmembrane protein 1) is a member of the IFN-inducible transmembrane protein family. It is an essential mediator of interferon-gamma-induced antiproliferation and plays a role in the control of cell growth. IFITM1 is upregulated in several tumor types and may be useful as a tumor biomarker. Both mouse IFITM1 and IFITM3 are expressed on the cell surfaces of primordial germ cells in a developmentally-regulated manner. IFITM1 activity is required for primordial germ cell transit, and IFITM1 acts as a repulsive molecule by repelling non-IFITM1-expressing primordial germ cells from the mesoderm into the endoderm.

Specifications

Antigen IFITM1
Applications Immunohistochemistry (Paraffin), Western Blot, Flow Cytometry, Immunoprecipitation
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene IFITM1
Gene Accession No. P13164
Gene Alias 9 27, CD225, IFI17, IFITM1, Interferon induced protein 17, Leu 13 antigen, LEU13
Gene Symbols IFITM1
Host Species Rabbit
Immunogen IFITM1 Fusion Protein Ag2320
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293618, 8519
Target Species Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.