Learn More
IFIT3 Mouse anti-Human, Unconjugated, Clone: 2C8B10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67208-1-IG-20UL
 View more versions of this product
Product Code. 16849725
  • 1710.33 NOK / 20µL
Description
The IFIT3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IFIT3. This antibody recognizes human antigen. The IFIT3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
IFIT3
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
2C8B10
Unconjugated
20 μL
3437
IgG2a