Learn More
IFIT3 Mouse anti-Human, Unconjugated, Clone: 2C8B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67208-1-IG-20UL
This item is not returnable.
View return policy
Description
The IFIT3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IFIT3. This antibody recognizes human antigen. The IFIT3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| IFIT3 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 2C8B10 | |
| Unconjugated | |
| 20 μL | |
| 3437 | |
| IgG2a |