missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFI27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568038
This item is not returnable.
View return policy
Description
IFI27 Polyclonal Antibody for IHC, Western Blot
Promotes cell death. Mediates IFN-induced apoptosis characterized by a rapid and robust release of cytochrome C from the mitochondria and activation of BAX and caspases 2, 3, 6, 8 and 9.
Specifications
| IFI27 | |
| Polyclonal | |
| Unconjugated | |
| IFI27 | |
| 2310061N23Rik; FAM14D; IFI27; interferon alpha inducible protein 27; interferon alpha-induced 11.5 kDa protein; Interferon alpha-inducible protein 27, mitochondrial; interferon, alpha-inducible protein 27; interferon-stimulated gene 12a protein; ISG12; ISG12(a); ISG12A; p27 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3429 | |
| -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P40305 | |
| IFI27 | |
| Synthesized peptide derived from human IFI27 (aa 1-21). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction