Learn More
IEICO-4F, Thermo Scientific Chemicals
Product Code. 17156554
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
Product Code. 17156554
Brand: Thermo Scientific Alfa Aesar H66546.MC
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2089044-02-8
Molecular Formula C103H100F4N4O4S4
Molecular Weight (g/mol) 1662.19
InChI Key JNJCNYUQCFAGFR-UHFFFAOYNA-N
Synonym 2,2 '-((2 Z,2 'Z)-(((4,4,9,9-tetrakis(4-hexylphenyl)-4,9-dihydro-sindaceno[1,2-b:5,6-b']dithiophene-2,7-diyl)bis(4-((2-ethylhexyl)oxy)thiophene-5,2- diyl))bis(methanylylidene))bis(5,6-difluoro-3-oxo-2,3-dihydro-1 H-indene-2,1- diylidene))dimalononitrile
IUPAC Name 2-[2-({5-[15-(5-{[1-(dicyanomethylidene)-5,6-difluoro-3-oxo-2,3-dihydro-1H-inden-2-ylidene]methyl}-3-[(2-ethylhexyl)oxy]thiophen-2-yl)-9,9,18-tris(4-hexylphenyl)-18-methyl-5,14-dithiapentacyclo[10.6.0.0³,¹⁰.0⁴,⁸.0¹³,¹⁷]octadeca-1(12),2,4(8),6,10,13(17),15-heptaen-6-yl]-4-[(2-ethylhexyl)oxy]thiophen-2-yl}methylidene)-5,6-difluoro-3-oxo-2,3-dihydro-1H-inden-1-ylidene]propanedinitrile
SMILES CCCCCCC1=CC=C(C=C1)C1(C)C2=C(SC(=C2)C2=C(OCC(CC)CCCC)C=C(S2)C=C2C(=O)C3=CC(F)=C(F)C=C3C2=C(C#N)C#N)C2=C1C=C1C3=C(C=C(S3)C3=C(OCC(CC)CCCC)C=C(S3)C=C3C(=O)C4=CC(F)=C(F)C=C4C3=C(C#N)C#N)C(C1=C2)(C1=CC=C(CCCCCC)C=C1)C1=CC=C(CCCCCC)C=C1
Specifications
Quantity 100 mg
Formula Weight 1808.40
Physical Form Solid
Chemical Name or Material IEICO-4 F