missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDUA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55158-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to IDUA.
IDUA encodes an enzyme that hydrolyzes the terminal alpha-L-iduronic acid residues of two glycosaminoglycans, dermatan sulfate and heparan sulfate. This hydrolysis is required for the lysosomal degradation of these glycosaminoglycans.Specifications
| IDUA | |
| Polyclonal | |
| Unconjugated | |
| IDUA | |
| Alpha L iduronidase, IDA, IDUA, iduronidase, alpha L, MPS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15932, 3425, 360904 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35475, P48441 | |
| IDUA | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction