missing translation for 'onlineSavingsMsg'
Learn More

IDO5L, MedChemExpress

Product Code. p-200097743
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16491776

Brand: Medchem Express HY15683

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IDO5L is a potent indoleamine 2,3-dioxygenase (IDO) inhibitor with an IC50 of 67 nM.

IDO5L is a potent indoleamine 2,3-dioxygenase (IDO) inhibitor with an IC50 of 67 nM.

Specifications

Chemical Name or Material IDO5L
CAS 914471-09-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C9H7ClFN5O2
Quantity 500 mg
Solubility Information DMSO : ≥ 52 mg/mL (191.43 mM)
SMILES ON/C(C1=NON=C1N)=N/C2=CC=C(F)C(Cl)=C2
Molecular Weight (g/mol) 271.64
Formula Weight 271.64
Percent Purity 95.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.