Learn More
IDO-IN-7, MedChemExpress
Product Code. 16476376
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16476376
Brand: Medchem Express HY13983
MedChemExpress IDO-IN-7 (NLG-919 analogue) is a a potent IDO1 inhibitor with an IC50 of 38 nM.
IDO-IN-7 (NLG-919 analogue) is a a potent IDO1 inhibitor with an IC50 of 38 nM.
Specifications
Chemical Name or Material IDO-IN-7
CAS 1402836-58-1
Color White
Purity Grade Notes Research
Molecular Formula C18H22N2O
Quantity 100 mg
Synonym NLG-919 analogue GDC-0919 analogue
Solubility Information DMSO : 50 mg/mL (177.07 mM; Need ultrasonic) ∣Ethanol : 25 mg/mL (88.53 mM; Need ultrasonic)
SMILES OC(C1CCCCC1)CC(C2=C3C=CC=C2)N4C3=CN=C4
Molecular Weight (g/mol) 282.38
Formula Weight 282.38
Percent Purity 96.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.