Learn More
IDO-IN-1, MedChemExpress
Product Code. 16498238
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498238 10 mg 10mg
16480688 5 mg 5mg
16474127 50 mg 50mg
16430366 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16498238 Supplier Medchem Express Supplier No. HY79531
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
MedChemExpress IDO-IN-1 is a potent indoleamine 2,3-dioxygenase (IDO) inhibitor with an IC50 of 59 nM.
IDO-IN-1 is a potent indoleamine 2,3-dioxygenase (IDO) inhibitor with an IC50 of 59 nM.
Specifications
Chemical Name or Material IDO-IN-1
CAS 914638-30-5
Color Gray
Purity Grade Notes Research
Molecular Formula C9H7BrFN5O2
Quantity 10 mg
Solubility Information DMSO : 120 mg/mL (379.64 mM; Need ultrasonic)
SMILES NC1=NON=C1/C(NO)=N/C2=CC(Br)=C(F)C=C2
Molecular Weight (g/mol) 316.09
Formula Weight 316.09
Percent Purity 95.3%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death