missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDE Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21728-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to IDE.
This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.Specifications
IDE | |
Polyclonal | |
Unconjugated | |
IDE | |
Abeta degrading protease, IDE, insulin degrading enzyme, Insulin protease, Insulinase, INSULYSIN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3416 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P14735 | |
IDE | |
IDE Fusion Protein Ag16392 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |