missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ ICP internal standard - 6 components: 100mg/l each of Bi; In; 6Li; Sc; Tb; Y in HNO3 5%

Product Code. 31034062 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 31034062

Brand: Fisherbrand™ EQ 0022 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Certified Reference Materials (CRMs) are used in ICP Mass Spectroscopy to ensure accuracy, traceability, and reliability of your measurements. They act as calibration benchmarks, guaranteeing that your instrument produces results that are scientifically valid and internationally comparable.

  • 6 components: 100 mg/L each of Bi ; In ; 6Li ; Sc ; Tb ; Y
  • Equivalent to Agilent ref 6610030100
  • Certified Reference Material (CRM) according to ISO 17025 and ISO 17034.
  • Each CRM comes with a certificate of analysis that includes uncertainty calculations.
  • This documentation is critical for audits, method validation, and proving the reliability of your data.

Specifications

Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.